Methyl-2-bromo-6-methylbenzoate
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR17046-1G
OR17046-5G
APOSOR17046-1GEA
162
DKK
APOSOR17046-1G
APOSOR17046-5G
Methyl-2-bromo-6-methylbenzoate
Methyl-2-bromo-6-methylbenzoate
Formel:
C₉H₉BrO₂ Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD09260938 CAS-nummer: 99548-56-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...