Dibromoisocyanuric acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: DBI, 1,3-Dibromo-1,3,5-triazine-2,4,6-trione

OR16500-25G OR16500-5G OR16500-1G
APOSOR16500-25GEA 1830 DKK
APOSOR16500-25G APOSOR16500-5G APOSOR16500-1G
Dibromoisocyanuric acid
Dibromoisocyanuric acid
Formel: C₃HBr₂N₃O₃
Smeltepunkt: 309 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00463941
CAS-nummer: 15114-43-9
UN nummer: 3085
ADR: 5.1+8,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR