Tetrabutylammoniumnitrat

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

OR16429-100G OR16429-500G
APOSOR16429-100GEA 1570 DKK
APOSOR16429-100G APOSOR16429-500G
Tetrabutylammoniumnitrat
Tetrabutylammonium nitrate
Formel: C₁₆H₃₆N₂O₃
Smeltepunkt: 116…119 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00043202
CAS-nummer: 1941-27-1
EINECS: 217-726-0
UN nummer: 1479
ADR: 5.1,III

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR