Ethyl thieno[2,3-b]pyridine-2-carboxylate
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR15760-1G
OR15760-5G
APOSOR15760-1GEA
654
DKK
APOSOR15760-1G
APOSOR15760-5G
Ethyl thieno[2,3-b]pyridine-2-carboxylate
Ethyl thieno[2,3-b]pyridine-2-carboxylate
Formel:
C₁₀H₉NO₂S Smeltepunkt: 61…63 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD07772793 CAS-nummer: 59944-78-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...