5-Methoxy-3-pyridineboronic acid pinacol ester 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 5-Methoxy-3-pyridylboronic acid Pinacol ester, 3-Methoxypyridine-5-boronic acid Pinacol ester, 3-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-(5-Methoxy-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR15595-25G OR15595-5G
APOSOR15595-25GEA 2050 DKK
APOSOR15595-25G APOSOR15595-5G
5-Methoxy-3-pyridineboronic acid pinacol ester 97%
5-Methoxy-3-pyridineboronic acid pinacol ester
Formel: C₁₂H₁₈BNO₃
Molekylvægt: 235.0872 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03412068
CAS-nummer: 445264-60-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR