5-Amino-3-phenylisoxazol 95+%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR15239-1G
OR15239-5G
APOSOR15239-1GEA
447
DKK
APOSOR15239-1G
APOSOR15239-5G
5-Amino-3-phenylisoxazol 95+%
5-Amino-3-phenylisoxazole
Formel:
C₉H₇NO Smeltepunkt: 109…114 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00464114 CAS-nummer: 4369-55-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...