tert-Butyl-3-(3-cyanophenyl)-2,2-dioxo-2λ~6~-diazathiane-1-carboxylate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 3-(3-Cyanophenyl)-2,2-dioxodiazathiane, N1-BOC protected

OR14819-10G OR14819-1G OR14819-5G
APOSOR14819-10GEA 6710 DKK
APOSOR14819-10G APOSOR14819-1G APOSOR14819-5G
tert-Butyl-3-(3-cyanophenyl)-2,2-dioxo-2λ~6~-diazathiane-1-carboxylate
tert-Butyl-3-(3-cyanophenyl)-2,2-dioxo-2lambda~6~-diazathiane-1-carboxylate
Formel: C₁₂H₁₅N₃O₄S
Molekylvægt: 297.34 g/mol
Smeltepunkt: 140…141 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09865021
CAS-nummer: 1017782-66-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR