(2R)-2-Amino-3-(pyridin-3-yl)propanoic acid 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR14724-1G
OR14724-250MG
OR14724-5G
APOSOR14724-1GEA
377
DKK
APOSOR14724-1G
APOSOR14724-250MG
APOSOR14724-5G
(2R)-2-Amino-3-(pyridin-3-yl)propanoic acid 98%
(2R)-2-Amino-3-(pyridin-3-yl)propanoic acid
Formel:
C₈H₁₀N₂O₂ Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00079673 CAS-nummer: 70702-47-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...