2-Hydroxy-6-methoxybenzoesyre
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-Hydroxy-6-methoxybenzoic acid, 6-Hydroxy-o-anisic acid
OR14337-1G
OR14337-5G
APOSOR14337-1GEA
164
DKK
APOSOR14337-1G
APOSOR14337-5G
2-Hydroxy-6-methoxybenzoesyre
2-Hydroxy-6-methoxybenzoesyre
Formel:
C₈H₈O₄ Molekylvægt: 168.15 g/mol Smeltepunkt: 134…138 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00674090 CAS-nummer: 3147-64-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...