2-Hydroxy-5-methoxybenzoesyre 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-Hydroxy-5-methoxybenzoic acid
OR14336-100G
OR14336-250G
OR14336-25G
APOSOR14336-100GEA
898
DKK
APOSOR14336-100G
APOSOR14336-250G
APOSOR14336-25G
2-Hydroxy-5-methoxybenzoesyre 98%
2-Hydroxy-5-methoxybenzoesyre
Formel:
C₈H₈O₄ Molekylvægt: 168.15 g/mol Kogepunkt: 327 Smeltepunkt: 141…143 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00002459 CAS-nummer: 2612-02-4 EINECS: 220-037-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...