4-Isopropoxyphenylboronic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR14157-100G OR14157-25G OR14157-5G
APOSOR14157-100GEA 1140 DKK
APOSOR14157-100G APOSOR14157-25G APOSOR14157-5G
4-Isopropoxyphenylboronic acid
4-Isopropoxyphenylboronic acid
Formel: C₉H₁₃BO₃
Smeltepunkt: 150…154 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD03427051
CAS-nummer: 153624-46-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR