Methyl-3-amino-4-hydroxybenzoate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methyl 3-amino-4-hydroxybenzoate

OR14130-10G OR14130-1G OR14130-5G
APOSOR14130-10GEA 806 DKK
APOSOR14130-10G APOSOR14130-1G APOSOR14130-5G
Methyl-3-amino-4-hydroxybenzoate
Methyl-3-amino-4-hydroxybenzoate
Formel: C₈H₉NO₃
Molekylvægt: 167.164 g/mol
Smeltepunkt: 141…143 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06659667
CAS-nummer: 536-25-4
EINECS: 208-627-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR