Methyl-7-bromobenzo[b]thiophene-2-carboxylate
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR14066-1G
OR14066-5G
OR14066-10G
APOSOR14066-1GEA
349
DKK
APOSOR14066-1G
APOSOR14066-5G
APOSOR14066-10G
Methyl-7-bromobenzo[b]thiophene-2-carboxylate
Methyl-7-bromobenzo[b]thiophene-2-carboxylate
Formel:
C₁₀H₇BrO₂S Smeltepunkt: 99…102 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD09027095 CAS-nummer: 550998-53-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...