(±)-α-Methoxyphenyleddikesyre
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
DL-α-Methoxyphenylacetic acid, 2-Methoxy-2-phenylacetic acid, α-Methoxyphenylacetic acid
OR13894-1G
OR13894-500MG
OR13894-5G
APOSOR13894-1GEA
186
DKK
APOSOR13894-1G
APOSOR13894-500MG
APOSOR13894-5G
(±)-α-Methoxyphenyleddikesyre
(±)-α-Methoxyphenyleddikesyre
Formel:
C₉H₁₀O₃ Molekylvægt: 166.18 g/mol Kogepunkt: 165 °C/18mmHg Smeltepunkt: 69…71 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00064217 CAS-nummer: 7021-09-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...