tert-Butyl hydrogen malonate 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR13710-100G
OR13710-25G
OR13710-5G
APOSOR13710-100GEA
513
DKK
APOSOR13710-100G
APOSOR13710-25G
APOSOR13710-5G
tert-Butyl hydrogen malonate 95%
tert-Butyl hydrogen malonate
Formel:
C₇H₁₂O₄ Kogepunkt: 90 °C/2mmHg Smeltepunkt: 19…20 °C Massefylde: 1,04 g/cm³ (25 °C) Flammepunkt: 91 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00191886 CAS-nummer: 40052-13-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...