5-Amino-2-chlorpyridin 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR1348-25G
OR1348-5G
APOSOR1348-25GEA
186
DKK
APOSOR1348-25G
APOSOR1348-5G
5-Amino-2-chlorpyridin 98%
5-Amino-2-chlorpyridin
Formel:
C₅H₅ClN₂ Smeltepunkt: 80…84 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00006243 CAS-nummer: 5350-93-6 EINECS: 226-322-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...