2-Cyanobenzaldehyd

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-Formylbenzonitrile

OR13206-25G OR13206-50G OR13206-5G
APOSOR13206-25GEA 186 DKK
APOSOR13206-25G APOSOR13206-50G APOSOR13206-5G
2-Cyanobenzaldehyd
2-Cyanobenzaldehyd
Formel: C₈H₅NO
Smeltepunkt: 103…105 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00017503
CAS-nummer: 7468-67-9
UN nummer: 3439
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR