4-Brombenzylbromid 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
4-Bromobenzyl bromide, 1-Bromo-4-(bromomethyl)benzene
OR13169-100G
OR13169-25G
OR13169-500G
APOSOR13169-100GEA
818
DKK
APOSOR13169-100G
APOSOR13169-25G
APOSOR13169-500G
4-Brombenzylbromid 98%
4-Brombenzylbromid
Formel:
C₇H₆Br₂ Molekylvægt: 249.93 g/mol Kogepunkt: 115…124 °C/12mmHg Smeltepunkt: 60…64 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000179 CAS-nummer: 589-15-1 EINECS: 209-636-5 UN nummer: 3261 ADR: 8 II Merck Index: 13,01393 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...