Cyclohexylboronic acid 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR13116-100G
OR13116-25G
OR13116-5G
APOSOR13116-100GEA
1910
DKK
APOSOR13116-100G
APOSOR13116-25G
APOSOR13116-5G
Cyclohexylboronic acid 97%
Cyclohexylboronic acid
Formel:
C₆H₁₃BO₂ Smeltepunkt: 122…123 °C Opbevaringstemperatur: Fryser |
MDL Number:
MFCD01311824 CAS-nummer: 4441-56-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...