Methyl-4-acetylbenzoate 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR13031-100G
OR13031-25G
OR13031-5G
APOSOR13031-100GEA
1160
DKK
APOSOR13031-100G
APOSOR13031-25G
APOSOR13031-5G
Methyl-4-acetylbenzoate 98%
Methyl-4-acetylbenzoate
Formel:
C₁₀H₁₀O₃ Smeltepunkt: 92…96 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00216474 CAS-nummer: 3609-53-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...