4-Methyl-1,2,3-thiadiazole-5-carboxylic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR1271-100G OR1271-25G
APOSOR1271-100GEA 5220 DKK
APOSOR1271-100G APOSOR1271-25G
4-Methyl-1,2,3-thiadiazole-5-carboxylic acid
4-Methyl-1,2,3-thiadiazole-5-carboxylic acid
Formel: C₄H₄N₂O₂S
Smeltepunkt: 166…169 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00173830
CAS-nummer: 18212-21-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR