5-Bromoindole-2-carboxylic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 5-Bromo-1H-indole-2-carboxylic acid

OR12042-1G OR12042-5G
APOSOR12042-1GEA 175 DKK
APOSOR12042-1G APOSOR12042-5G
5-Bromoindole-2-carboxylic acid
5-Bromoindole-2-carboxylic acid
Formel: C₉H₆BrNO₂
Molekylvægt: 240.06 g/mol
Smeltepunkt: 287…288 °C
Opbevaringstemperatur: Fryser
MDL Number: MFCD00022705
CAS-nummer: 7254-19-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR