4-Bromo-N,N-diethylbenzamide

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR11919-1G OR11919-5G OR11919-25G
APOSOR11919-1GEA 164 DKK
APOSOR11919-1G APOSOR11919-5G APOSOR11919-25G
4-Bromo-N,N-diethylbenzamide
4-Bromo-N,N-diethylbenzamide
Formel: C₁₁H₁₄BrNO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00463697
CAS-nummer: 5892-99-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR