2-Amino-5-nitropyridin

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 5-Nitropyridin-2-ylamine, 5-Nitropyridin-2-amine, 5-Nitro-2-pyridylamine

OR1164-100G OR1164-25G OR1164-500G
APOSOR1164-100GEA 218 DKK
APOSOR1164-100G APOSOR1164-25G APOSOR1164-500G
2-Amino-5-nitropyridin
2-Amino-5-nitropyridine
Formel: C₅H₅N₃O₂
Smeltepunkt: 186…190 °C
Flammepunkt: >110 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006325
CAS-nummer: 4214-76-0
EINECS: 224-145-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR