Ethyl 2-(tert-butoxycarbonylamino)oxazole-5-carboxylate 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR11529-1G OR11529-5G
APOSOR11529-1GEA 1860 DKK
APOSOR11529-1G APOSOR11529-5G
Ethyl 2-(tert-butoxycarbonylamino)oxazole-5-carboxylate 98%
Ethyl-2-(tert-butoxycarbonylamino)oxazole-5-carboxylate
Formel: C₁₁H₁₆N₂O₅
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09475909
CAS-nummer: 941294-50-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR