1-Bromo-2-(methoxymethyl)benzene 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR11434-1G OR11434-5G OR11434-25G
APOSOR11434-1GEA 164 DKK
APOSOR11434-1G APOSOR11434-5G APOSOR11434-25G
1-Bromo-2-(methoxymethyl)benzene 97%
1-Bromo-2-(methoxymethyl)benzene
Formel: C₈H₉BrO
Kogepunkt: 75…80 °C/0,5mmHg
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03790887
CAS-nummer: 52711-30-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR