6-Chloro-3-formylchromone

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 6-Chloro-4-oxo-4H-chromene-3-carbaldehyde

OR1141-250MG OR1141-500MG
APOSOR1141-250MGEA 656 DKK
APOSOR1141-250MG APOSOR1141-500MG
6-Chloro-3-formylchromone
6-Chloro-3-formylchromone
Formel: C₁₀H₅ClO₃
Molekylvægt: 208.597 g/mol
Smeltepunkt: 166…168 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00139138
CAS-nummer: 42248-31-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR