4-Hydroxy-2-mercapto-6-propylpyrimidin 97+%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Propylthiouracil, 4-Hydroxy-2-mercapto-6-propylpyrimidine, 4-Propyl-2-thiouracil, 6-Propyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one
OR11311-100G
OR11311-25G
APOSOR11311-100GEA
0
DKK
APOSOR11311-100G
APOSOR11311-25G
4-Hydroxy-2-mercapto-6-propylpyrimidin 97+%
6-Propyl-2-thiouracil
Formel:
C₇H₁₀N₂OS Smeltepunkt: 218…220 °C Flammepunkt: 300 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00006041 CAS-nummer: 51-52-5 EINECS: 200-103-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...