Imidazo[1,2-a]pyridine-6-carbaldehyde 96%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR111513-1G OR111513-5G OR111513-250MG
APOSOR111513-1GEA 2130 DKK
APOSOR111513-1G APOSOR111513-5G APOSOR111513-250MG
Imidazo[1,2-a]pyridine-6-carbaldehyde 96%
Imidazo[1,2-a]pyridine-6-carbaldehyde
Formel: C₈H₆N₂O
Molekylvægt: 146.15 g/mol
Smeltepunkt: 151…153 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07772808
CAS-nummer: 116355-16-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR