2-[(2-Amino-6-chloropyrimidin-4-yl)amino]ethanol
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR111048-1G
OR111048-5G
APOSOR111048-1GEA
0
DKK
APOSOR111048-1G
APOSOR111048-5G
2-[(2-Amino-6-chloropyrimidin-4-yl)amino]ethanol
2-[(2-Amino-6-chloropyrimidin-4-yl)amino]ethanol
Formel:
C₆H₉ClN₄O Smeltepunkt: 150…151 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD13188631 CAS-nummer: 2846-77-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...