4-Methyl-2-phenyl-1,3-thiazole-5-carbonitrile

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR110680-1G OR110680-5G
APOSOR110680-1GEA 933 DKK
APOSOR110680-1G APOSOR110680-5G
4-Methyl-2-phenyl-1,3-thiazole-5-carbonitrile
4-Methyl-2-phenyl-1,3-thiazole-5-carbonitrile
Formel: C₁₁H₈N₂S
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD05664409
CAS-nummer: 830330-33-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR