1,14-Dibromtetradecan

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR10848-1G OR10848-5G OR10848-25G
APOSOR10848-1GEA 371 DKK
APOSOR10848-1G APOSOR10848-5G APOSOR10848-25G
1,14-Dibromtetradecan
1,14-Dibromotetradecane
Formel: BrH₂C(CH₂)₁₂CH₂Br
Kogepunkt: 190…192 °C/8mmHg
Smeltepunkt: 45…50 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00045049
CAS-nummer: 37688-96-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR