Tetrahydropyran-4-acetic acid 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Tetrahydropyranyl-4-acetic acid

OR10586-1G OR10586-5G OR10586-10G
APOSOR10586-1GEA 164 DKK
APOSOR10586-1G APOSOR10586-5G APOSOR10586-10G
Tetrahydropyran-4-acetic acid 97%
Tetrahydropyran-4-acetic acid
Formel: C₇H₁₂O₃
Molekylvægt: 144.1700 g/mol
Smeltepunkt: 60…63 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01631204
CAS-nummer: 85064-61-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR