4-Nitrophenylboronsyre
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Nitrobenzeneboronic acid, (4-Nitrophenyl)boronic acid, Dihydroxy(4-nitrophenyl)borane, (4-Nitrophenyl)metaboric acid
OR10572-1G
OR10572-5G
OR10572-25G
APOSOR10572-1GEA
164
DKK
APOSOR10572-1G
APOSOR10572-5G
APOSOR10572-25G
4-Nitrophenylboronsyre
4-Nitrophenylboronsyre
Formel:
C₆H₆BNO₄ Molekylvægt: 166.93 g/mol Smeltepunkt: 285…290 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00161360 CAS-nummer: 24067-17-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...