(2-Methylsulphinylphenyl)boronic acid 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR10558-100MG
OR10558-1G
APOSOR10558-100MGEA
545
DKK
APOSOR10558-100MG
APOSOR10558-1G
(2-Methylsulphinylphenyl)boronic acid 95%
(2-Methylsulphinylphenyl)boronic acid
Formel:
C₇H₉BO₃S Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD03095261 CAS-nummer: 850567-97-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...