3-(Hydroxymethyl)phenylboronic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR10500-100G OR10500-25G OR10500-5G
APOSOR10500-100GEA 1480 DKK
APOSOR10500-100G APOSOR10500-25G APOSOR10500-5G
3-(Hydroxymethyl)phenylboronic acid
3-(Hydroxymethyl)phenylboronic acid
Formel: C₇H₉BO₃
Smeltepunkt: 93…99 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01317846
CAS-nummer: 87199-15-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR