5-Bromoxindol
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
5-Bromo-1,3-dihydro-2H-indol-2-one, 5-Bromoindolin-2-one, 5-Bromo-2-oxindole
OR10383-100G
OR10383-25G
OR10383-5G
APOSOR10383-100GEA
1590
DKK
APOSOR10383-100G
APOSOR10383-25G
APOSOR10383-5G
5-Bromoxindol
5-Bromoxindol
Formel:
C₈H₆BrNO Smeltepunkt: 220…224 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00456998 CAS-nummer: 20870-78-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...