3-Benzyl-3H-imidazole-4-carbaldehyde 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR10353-10G
OR10353-1G
OR10353-5G
APOSOR10353-10GEA
6800
DKK
APOSOR10353-10G
APOSOR10353-1G
APOSOR10353-5G
3-Benzyl-3H-imidazole-4-carbaldehyde 98%
3-Benzyl-3H-imidazole-4-carbaldehyde
Formel:
C₁₁H₁₀N₂O Smeltepunkt: 46…49 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD02179539 CAS-nummer: 85102-99-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...