4-Amino-2-chlorpyridin

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR10317-100G OR10317-500G
APOSOR10317-100GEA 458 DKK
APOSOR10317-100G APOSOR10317-500G
4-Amino-2-chlorpyridin
4-Amino-2-chloropyridine
Formel: C₅H₅ClN₂
Smeltepunkt: 90…94 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00060089
CAS-nummer: 14432-12-3
EINECS: 238-403-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR