2-Chlorethylammoniumchlorid 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-Chloroethylamine hydrochloride, 2-Chloroethylamine HCl, 2-Chloroethanamine hydrochloride

OR0986-100G OR0986-25G
APOSOR0986-100GEA 284 DKK
APOSOR0986-100G APOSOR0986-25G
2-Chlorethylammoniumchlorid 97%
2-Chlorethylammoniumchlorid
Formel: C₂H₇Cl₂N
Molekylvægt: 115.99 g/mol
Smeltepunkt: 140…150 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00012887
CAS-nummer: 870-24-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR