2,6-Dihydroxy-9,10-anthraquinon 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2,6-Dihydroxyanthracene-9,10-dione, Anthraflavic acid, 2,6-Dihydroxyanthraquinone
OR0392-25G
OR0392-5G
APOSOR0392-25GEA
1810
DKK
APOSOR0392-25G
APOSOR0392-5G
2,6-Dihydroxy-9,10-anthraquinon 97%
2,6-Dihydroxy-9,10-anthraquinon
Formel:
C₁₄H₈O₄ Smeltepunkt: >320 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001228 CAS-nummer: 84-60-6 EINECS: 201-544-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...