Ethyl-3-bromoimidazo[1,2-a]pyridine-2-carboxylate
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Ethyl 3-bromoimidazo[1,2-a]pyridine-2-carboxylate
OR01982-500MG
OR01982-1G
OR01982-5G
APOSOR01982-500MGEA
186
DKK
APOSOR01982-500MG
APOSOR01982-1G
APOSOR01982-5G
Ethyl-3-bromoimidazo[1,2-a]pyridine-2-carboxylate
Ethyl-3-bromoimidazo[1,2-a]pyridine-2-carboxylate
Formel:
C₁₀H₉BrN₂O₂ Smeltepunkt: 76…79 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD05864806 CAS-nummer: 143982-54-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...