3,4-Dihydro-1H-benzo[e][1,4]diazepine-2,5-dione
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR01717-1G
OR01717-5G
APOSOR01717-1GEA
1010
DKK
APOSOR01717-1G
APOSOR01717-5G
3,4-Dihydro-1H-benzo[e][1,4]diazepine-2,5-dione
3,4-Dihydro-1H-benzo[e][1,4]diazepine-2,5-dione
Formel:
C₉H₈N₂O₂ Smeltepunkt: 327…328 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00111349 CAS-nummer: 5118-94-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...