Methyl(4-aminophenyl)acetate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methyl-(4-aminophenyl)acetate, Methyl 2-(4-aminophenyl)acetate

OR0171-1G OR0171-5G
APOSOR0171-1GEA 164 DKK
APOSOR0171-1G APOSOR0171-5G
Methyl(4-aminophenyl)acetate
Methyl(4-aminophenyl)acetate
Formel: C₉H₁₁NO₂
Molekylvægt: 165.19 g/mol
Kogepunkt: 96…100 °C/0.1mmHg
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06200783
CAS-nummer: 39552-81-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR