Ethyl 2-naphthoyl formate, Technical Grade
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Ethyl (naphth-2-yl)(oxo)acetate, Ethyl 2-(naphthalen-2-yl)-2-oxoacetate
OR0142-250MG
OR0142-1G
APOSOR0142-250MGEA
1490
DKK
APOSOR0142-250MG
APOSOR0142-1G
Ethyl 2-naphthoyl formate, Technical Grade
Ethyl-2-naphthoyl formate
Formel:
C₁₄H₁₂O₃ Kogepunkt: 138…140 °C/0,15mmHg Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01934866 CAS-nummer: 73790-09-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...