2-Chlorkanelsyre
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR0066-250MG
OR0066-500MG
OR0066-1G
OR0066-2.5G
OR0066-5G
APOSOR0066-250MGEA
437
DKK
APOSOR0066-250MG
APOSOR0066-500MG
APOSOR0066-1G
APOSOR0066-2.5G
APOSOR0066-5G
2-Chlorkanelsyre
2-Chlorkanelsyre
Formel:
C₉H₇ClO₂ Molekylvægt: 182.61 g/mol Kogepunkt: 311 Smeltepunkt: 209…212 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00004372 CAS-nummer: 3752-25-8 EINECS: 223-154-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...