Thiophene-2-carboxamide

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR0041-25G OR0041-5G
APOSOR0041-25GEA 1020 DKK
APOSOR0041-25G APOSOR0041-5G
Thiophene-2-carboxamide
Thiophene-2-carboxamide
Formel: C₅H₅NOS
Smeltepunkt: 181…183 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010425
CAS-nummer: 5813-89-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR