Yttrium(III)chlorid, anhydrous 99,9%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
IN3844-25G
IN3844-5G
APOSIN3844-25GEA
1600
DKK
APOSIN3844-25G
APOSIN3844-5G
Yttrium(III)chlorid, anhydrous 99,9%
Yttrium(III) chloride
Formel:
Cl₃Y Kogepunkt: 1507 °C Smeltepunkt: 721 °C Massefylde: 2,67 g/cm³ (25 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011470 CAS-nummer: 10361-92-9 EINECS: 233-801-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...