Thulium(III)oxid 99.99%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

IN3616-1G IN3616-5G
APOSIN3616-1GEA 933 DKK
APOSIN3616-1G APOSIN3616-5G
Thulium(III)oxid 99.99%
Thulium(III) oxide
Formel: O₃Tm₂
Molekylvægt: 385.87 g/mol
Smeltepunkt: 2425 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011285
CAS-nummer: 12036-44-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR