Dysprosium(III)oxid 99,9%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Didysprosium trioxide, Dysprosium oxide

IN1663-100G IN1663-250G IN1663-25G
APOSIN1663-100GEA 4640 DKK
APOSIN1663-100G APOSIN1663-250G APOSIN1663-25G
Dysprosium(III)oxid 99,9%
Dysprosium(III) oxide
Formel: Dy₂O₃
Molekylvægt: 373 g/mol
Smeltepunkt: 2340 °C
Massefylde: 7.81 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010986
CAS-nummer: 1308-87-8
EINECS: 215-164-0
Merck Index: 14,03482

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR